Product Details

SNP ID
rs838131
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:48757420 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGGACCTTGAGTGCTAGGGAGAC[A/C]CCCCGCCTGGCCTGAGAGAGCACTG
Phenotype
MIM: 609436 MIM: 211100 MIM: 609278
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FGF21 PubMed Links

Gene Details

Gene
FGF21
Gene Name
fibroblast growth factor 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019113.3 Intron NP_061986.1
Gene
FUT1
Gene Name
fucosyltransferase 1 (H blood group)
There are no transcripts associated with this gene.

Gene
IZUMO1
Gene Name
izumo sperm-egg fusion 1
There are no transcripts associated with this gene.

View Full Product Details