Product Details

SNP ID
rs1269946
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:59458171 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCCTACAGTCACCCTTGGAGAACA[A/G]TAGCTTTAGTAGGGCTCTGTTTGAC
Phenotype
MIM: 607970 MIM: 614811
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPR135 PubMed Links

Gene Details

Gene
GPR135
Gene Name
G protein-coupled receptor 135
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022571.5 Intron NP_072093.2
XM_017021596.1 Intron XP_016877085.1
XM_017021597.1 Intron XP_016877086.1
XM_017021598.1 Intron XP_016877087.1
XM_017021599.1 Intron XP_016877088.1
Gene
L3HYPDH
Gene Name
trans-L-3-hydroxyproline dehydratase
There are no transcripts associated with this gene.

View Full Product Details