Product Details

SNP ID
rs3807308
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:128831678 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGGGGCTTCCGCAAGTCCAGGCC[C/T]TGCCCCAGCCCACCCCCTACCGCGG
Phenotype
MIM: 611902 MIM: 102565
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC136 PubMed Links

Gene Details

Gene
CCDC136
Gene Name
coiled-coil domain containing 136
There are no transcripts associated with this gene.

Gene
FLNC
Gene Name
filamin C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127487.1 Intron NP_001120959.1
NM_001458.4 Intron NP_001449.3

View Full Product Details