Product Details

SNP ID
rs1151623
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:63737253 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGGCTATGGTGGCCACAGCTGTC[C/T]TGCCCCTCCTCACGGCCGTGCTGGG
Phenotype
MIM: 609809 MIM: 609493 MIM: 614639
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LIME1 PubMed Links

Gene Details

Gene
LIME1
Gene Name
Lck interacting transmembrane adaptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001305654.1 Intron NP_001292583.1
NM_001305655.1 Intron NP_001292584.1
NM_017806.3 Intron NP_060276.2
Gene
SLC2A4RG
Gene Name
SLC2A4 regulator
There are no transcripts associated with this gene.

Gene
ZBTB46
Gene Name
zinc finger and BTB domain containing 46
There are no transcripts associated with this gene.

Gene
ZGPAT
Gene Name
zinc finger CCCH-type and G-patch domain containing
There are no transcripts associated with this gene.

View Full Product Details