Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001243259.1 | 69 | Intron | NP_001230188.1 | ||
NM_015517.4 | 69 | Intron | NP_056332.2 | ||
NM_198971.2 | 69 | Intron | NP_945322.1 | ||
XM_011542744.2 | 69 | Intron | XP_011541046.1 | ||
XM_011542745.2 | 69 | Intron | XP_011541047.1 | ||
XM_011542746.1 | 69 | Intron | XP_011541048.1 | ||
XM_017017499.1 | 69 | Intron | XP_016872988.1 | ||
XM_017017500.1 | 69 | Intron | XP_016872989.1 | ||
XM_017017501.1 | 69 | Intron | XP_016872990.1 | ||
XM_017017502.1 | 69 | UTR 5 | XP_016872991.1 | ||
XM_017017503.1 | 69 | Intron | XP_016872992.1 | ||
XM_017017504.1 | 69 | Intron | XP_016872993.1 | ||
XM_017017505.1 | 69 | Intron | XP_016872994.1 | ||
XM_017017506.1 | 69 | Intron | XP_016872995.1 | ||
XM_017017507.1 | 69 | Intron | XP_016872996.1 | ||
XM_017017508.1 | 69 | Intron | XP_016872997.1 | ||
XM_017017509.1 | 69 | Intron | XP_016872998.1 |