Product Details

SNP ID
rs1059773
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.9:133361989 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GAGAACCCATGTCCTGAAGCCATTG[A/C]TCGATCTTACCAATATTCGGGGGTA
Phenotype
MIM: 185620 MIM: 185630 MIM: 185660
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
SURF1 PubMed Links
Additional Information
For this assay, SNP(s) [rs41309956] are located under a probe and SNP(s) [rs146867534] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
SURF1
Gene Name
surfeit 1
There are no transcripts associated with this gene.

Gene
SURF2
Gene Name
surfeit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278928.1 2511 Intron NP_001265857.1
NM_017503.4 2511 Intron NP_059973.4
Gene
SURF4
Gene Name
surfeit 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001280788.1 2511 UTR 3 NP_001267717.1
NM_001280789.1 2511 UTR 3 NP_001267718.1
NM_001280790.1 2511 UTR 3 NP_001267719.1
NM_001280791.1 2511 UTR 3 NP_001267720.1
NM_001280792.1 2511 UTR 3 NP_001267721.1
NM_001280793.1 2511 UTR 3 NP_001267722.1
NM_033161.3 2511 UTR 3 NP_149351.1
XM_011518943.1 2511 UTR 3 XP_011517245.1
XM_011518944.2 2511 UTR 3 XP_011517246.1
XM_011518945.2 2511 UTR 3 XP_011517247.1
XM_017015062.1 2511 UTR 3 XP_016870551.1

View Full Product Details