Product Details
- SNP ID
-
rs681475
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
76
- Location
-
Chr.1:70415034 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- ATTGCTCCCTATGCTTTGAGCCACT[C/T]TTGTAGAGCCAAGATGTGAATGAAT
- Phenotype
-
MIM: 607657
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CTH
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs115228206] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CTH
- Gene Name
- cystathionine gamma-lyase
View Full Product Details