Product Details

SNP ID
rs4910638
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:4599631 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGGCAAGACTAGGACACTGGTCT[A/C]CTGGAGTCCAAGGGACAGTGATTTG
Phenotype
MIM: 613184
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OR52I1 PubMed Links

Gene Details

Gene
OR52I1
Gene Name
olfactory receptor family 52 subfamily I member 1
There are no transcripts associated with this gene.

Gene
TRIM68
Gene Name
tripartite motif containing 68
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304496.1 2393 UTR 3 NP_001291425.1
NM_018073.7 2393 UTR 3 NP_060543.5

View Full Product Details