Product Details

SNP ID
rs1052656
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:34087362 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCAGAGAGGATTCTCCTCAAAGCCA[C/T]GGGGGCTATGGATGTGAATCTTAGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DCAF12 PubMed Links

Gene Details

Gene
DCAF12
Gene Name
DDB1 and CUL4 associated factor 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015397.3 2519 UTR 3 NP_056212.1
XM_011517843.1 2519 UTR 3 XP_011516145.1

View Full Product Details