Product Details

SNP ID
rs34466907
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:36326221 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATAAAACACTTAATGAAAAATTT[G/T]CAAATTGCTTTTCCTCTCCCTTTAA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KIAA0895 PubMed Links

Gene Details

Gene
KIAA0895
Gene Name
KIAA0895
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100425.1 2153 UTR 3 NP_001093895.1
NM_001199706.1 2153 UTR 3 NP_001186635.1
NM_001199707.1 2153 UTR 3 NP_001186636.1
NM_001199708.1 2153 UTR 3 NP_001186637.1
NM_001300956.1 2153 UTR 3 NP_001287885.1
NM_015314.2 2153 UTR 3 NP_056129.2
XM_005249689.3 2153 UTR 3 XP_005249746.1
XM_005249690.4 2153 UTR 3 XP_005249747.1

View Full Product Details