Product Details

SNP ID
rs10405636
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:18427932 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGACCTGTACTGCGCGGCGCCTGAC[A/C]GAAGAGAGGCCTGCGAGCACTCCGG
Phenotype
MIM: 611670 MIM: 607391
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ISYNA1 PubMed Links

Gene Details

Gene
ISYNA1
Gene Name
inositol-3-phosphate synthase 1
There are no transcripts associated with this gene.

Gene
SSBP4
Gene Name
single stranded DNA binding protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009998.3 578 Intron NP_001009998.1
NM_032627.4 578 Intron NP_116016.1
XM_005259790.3 578 Silent Mutation AGA,CGA R99R XP_005259847.1
XM_006722665.3 578 Silent Mutation AGA,CGA R99R XP_006722728.1
XM_006722666.2 578 Intron XP_006722729.1
XM_006722668.2 578 Intron XP_006722731.1
XM_017026431.1 578 Silent Mutation AGA,CGA R99R XP_016881920.1
XM_017026432.1 578 Silent Mutation AGA,CGA R88R XP_016881921.1
XM_017026433.1 578 Intron XP_016881922.1
XM_017026434.1 578 Silent Mutation AGA,CGA R99R XP_016881923.1
XM_017026435.1 578 Silent Mutation AGA,CGA R88R XP_016881924.1
XM_017026436.1 578 Intron XP_016881925.1
XM_017026437.1 578 Silent Mutation AGA,CGA R99R XP_016881926.1

View Full Product Details