Product Details

SNP ID
rs2059957
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:48183075 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCCACCACCCATAGCTTCTGGAC[A/G]GGGGAGCTGCATGGCACTGTGTACA
Phenotype
MIM: 613323
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FRMPD2 PubMed Links

Gene Details

Gene
FRMPD2
Gene Name
FERM and PDZ domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001018071.3 Intron NP_001018081.3
NM_001042512.2 Intron NP_001035977.2
NM_001318191.1 Intron NP_001305120.1
XM_011539327.2 Intron XP_011537629.1
XM_017015744.1 Intron XP_016871233.1

View Full Product Details