Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001161415.1 | 92 | UTR 5 | NP_001154887.1 | ||
NM_001161416.1 | 92 | UTR 5 | NP_001154888.1 | ||
NM_001161417.1 | 92 | UTR 5 | NP_001154889.1 | ||
NM_005291.2 | 92 | UTR 5 | NP_005282.1 | ||
XM_017003833.1 | 92 | Intron | XP_016859322.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001136037.2 | 92 | Intron | NP_001129509.2 | ||
NM_001161403.1 | 92 | Intron | NP_001154875.1 | ||
NM_001161404.1 | 92 | Intron | NP_001154876.1 | ||
NM_001256542.1 | 92 | Intron | NP_001243471.1 | ||
NM_017980.4 | 92 | Intron | NP_060450.2 | ||
XM_005263710.2 | 92 | Intron | XP_005263767.1 | ||
XM_006712627.3 | 92 | Intron | XP_006712690.1 | ||
XM_006712628.3 | 92 | Intron | XP_006712691.1 | ||
XM_011511453.1 | 92 | Intron | XP_011509755.1 | ||
XM_017004469.1 | 92 | Intron | XP_016859958.1 |