Product Details

SNP ID
rs4758441
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:6605445 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTTAATGAAGGGGAGGTAGTCAAC[A/G]GCAGTACAGGCTACTGAGAGATCAA
Phenotype
MIM: 602366 MIM: 615818 MIM: 600475 MIM: 607998
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ILK PubMed Links
Additional Information
For this assay, SNP(s) [rs151162142] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
ILK
Gene Name
integrin linked kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001014794.2 Intron NP_001014794.1
NM_001014795.2 Intron NP_001014795.1
NM_001278441.1 Intron NP_001265370.1
NM_001278442.1 Intron NP_001265371.1
NM_004517.3 Intron NP_004508.1
XM_005252904.4 Intron XP_005252961.1
XM_005252905.2 Intron XP_005252962.1
XM_011520065.1 Intron XP_011518367.1
XM_017017672.1 Intron XP_016873161.1
Gene
RRP8
Gene Name
ribosomal RNA processing 8, methyltransferase, homolog (yeast)
There are no transcripts associated with this gene.

Gene
TAF10
Gene Name
TATA-box binding protein associated factor 10
There are no transcripts associated with this gene.

Gene
TPP1
Gene Name
tripeptidyl peptidase 1
There are no transcripts associated with this gene.

View Full Product Details