Product Details

SNP ID
rs12414211
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:88808253 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTACTTTTGGATCATTGAGAATAT[A/G]GCCACAGAGAACTGAACCTAAAAGA
Phenotype
MIM: 613923
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LIPM PubMed Links

Gene Details

Gene
LIPM
Gene Name
lipase family member M
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128215.1 Intron NP_001121687.1
XM_011539748.2 Intron XP_011538050.1
XM_011539751.2 Intron XP_011538053.1
XM_011539752.2 Intron XP_011538054.1
XM_011539753.2 Intron XP_011538055.1

View Full Product Details