Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001319210.1 | Intron | NP_001306139.1 | |||
NM_001319211.1 | Intron | NP_001306140.1 | |||
NM_001319212.1 | Intron | NP_001306141.1 | |||
NM_003672.3 | Intron | NP_003663.2 | |||
NM_033312.2 | Intron | NP_201569.1 | |||
NM_033313.2 | Intron | NP_201570.1 | |||
XM_005271294.3 | Intron | XP_005271351.1 | |||
XM_005271296.3 | Intron | XP_005271353.1 | |||
XM_011542340.2 | Intron | XP_011540642.1 | |||
XM_011542341.2 | Intron | XP_011540643.1 | |||
XM_011542345.2 | Intron | XP_011540647.1 | |||
XM_011542346.2 | Intron | XP_011540648.1 | |||
XM_017002646.1 | Intron | XP_016858135.1 | |||
XM_017002647.1 | Intron | XP_016858136.1 |