Product Details

SNP ID
rs778585
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:140625792 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATGAATAAATTAATGACCAGAACA[C/T]TCCAGGCAGAGGGAAGAACAAGAGG
Phenotype
MIM: 158120
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CD14 PubMed Links

Gene Details

Gene
CD14
Gene Name
CD14 molecule
There are no transcripts associated with this gene.

Gene
TMCO6
Gene Name
transmembrane and coiled-coil domains 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300980.1 Intron NP_001287909.1
NM_001300982.1 Intron NP_001287911.1
NM_018502.4 Intron NP_060972.3
XM_005268477.1 Intron XP_005268534.1
XM_011537663.2 Intron XP_011535965.1
XM_011537665.2 Intron XP_011535967.1
XM_011537668.2 Intron XP_011535970.1
XM_017009617.1 Intron XP_016865106.1
XM_017009618.1 Intron XP_016865107.1
XM_017009619.1 Intron XP_016865108.1
XM_017009620.1 Intron XP_016865109.1

View Full Product Details