Product Details
- SNP ID
-
rs12466914
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:127050551 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CAGAGGATGCGGATCGCAAGTCAGA[C/G]CTTCCGTCCCACCTCCAGCCCTGCA
- Phenotype
-
MIM: 601248
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
BIN1
PubMed Links
Gene Details
- Gene
- BIN1
- Gene Name
- bridging integrator 1
View Full Product Details