Product Details
- SNP ID
-
rs329499
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:64096942 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGATGGAAATATGTGTATCATGTAG[G/T]TTTTTTCTCTTTAGAGATATTTAAA
- Phenotype
-
MIM: 614797
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
PELI1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74456017] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PELI1
- Gene Name
- pellino E3 ubiquitin protein ligase 1
View Full Product Details