Product Details

SNP ID
rs35388487
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:105610892 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCTTTTTTTTTTTTTTTTTTTGG[G/T]TGATTTTAATTTTAGCGCCATCGTC
Phenotype
MIM: 138246
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GRIA4 PubMed Links

Gene Details

Gene
GRIA4
Gene Name
glutamate ionotropic receptor AMPA type subunit 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000829.3 209 Intron NP_000820.3
NM_001077243.2 209 Intron NP_001070711.2
NM_001077244.1 209 Intron NP_001070712.1
NM_001112812.1 209 UTR 5 NP_001106283.1
XM_005271518.3 209 Intron XP_005271575.1
XM_006718823.1 209 Intron XP_006718886.1
XM_011542775.2 209 Intron XP_011541077.1
XM_011542776.2 209 Intron XP_011541078.1
XM_011542777.2 209 Intron XP_011541079.1
XM_017017609.1 209 Intron XP_016873098.1
XM_017017610.1 209 Intron XP_016873099.1
XM_017017611.1 209 Intron XP_016873100.1

View Full Product Details