Product Details

SNP ID
rs6928074
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:24680062 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAAGGGGGGTCCCTTATTAGTTGC[A/G]GAAGGAGTCGGGGGGATGCTGAGAT
Phenotype
MIM: 615652
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACOT13 PubMed Links

Gene Details

Gene
ACOT13
Gene Name
acyl-CoA thioesterase 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160094.1 Intron NP_001153566.1
NM_018473.3 Intron NP_060943.1

View Full Product Details