Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001127399.1 | Intron | NP_001120871.1 | |||
NM_001127400.1 | Intron | NP_001120872.1 | |||
NM_001127401.1 | Intron | NP_001120873.1 | |||
NM_016061.2 | Intron | NP_057145.1 | |||
XM_017004318.1 | Intron | XP_016859807.1 | |||
XM_017004319.1 | Intron | XP_016859808.1 | |||
XM_017004320.1 | Intron | XP_016859809.1 | |||
XM_017004321.1 | Intron | XP_016859810.1 | |||
XM_017004322.1 | Intron | XP_016859811.1 |