Product Details

SNP ID
rs7163748
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:57604113 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGGCTGGCCTGGTACTGTGAAGG[G/T]AAGGCTGGCCTTATCTCCTCTATGG
Phenotype
MIM: 614071
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GCOM1 PubMed Links

Gene Details

Gene
GCOM1
Gene Name
GRINL1A complex locus 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001018090.6 Intron NP_001018100.1
NM_001018091.6 Intron NP_001018101.1
NM_001285900.3 Intron NP_001272829.1
Gene
MYZAP
Gene Name
myocardial zonula adherens protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001018100.4 Intron NP_001018110.1
NM_152451.7 Intron NP_689664.3

View Full Product Details