Product Details

SNP ID
rs4359426
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:57358821 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGAGACATACAGGACAGAGCATGG[A/C]TCGCCTACAGACTGCACTCCTGGTT
Phenotype
MIM: 602957
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
CCL22 PubMed Links

Gene Details

Gene
CCL22
Gene Name
C-C motif chemokine ligand 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002990.4 59 Missense Mutation GAT,GCT D2A NP_002981.2
XM_017023531.1 59 Missense Mutation GAT,GCT D2A XP_016879020.1

View Full Product Details