Product Details

SNP ID
rs12126822
Assay Type
Functionally tested
NCBI dbSNP Submissions
17
Location
Chr.1:185066848 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTGGGGGCAGGGGTAGTAGTAGT[C/G]TTAGAGGAGATGAGATAGTGATTGA
Phenotype
MIM: 608985
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RNF2 PubMed Links

Gene Details

Gene
RNF2
Gene Name
ring finger protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007212.3 Intron NP_009143.1
XM_005245413.3 Intron XP_005245470.1
XM_011509851.2 Intron XP_011508153.1
XM_011509852.2 Intron XP_011508154.1

View Full Product Details