Product Details

SNP ID
rs36031159
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:54645251 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCCCACAGCTTTTTTTGCTCCGT[C/T]TAGGCCTTTTTCTAGGGTGGATTCA
Phenotype
MIM: 606634
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DCD PubMed Links

Gene Details

Gene
DCD
Gene Name
dermcidin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300854.1 401 Missense Mutation AAC,GAC N71D NP_001287783.1
NM_053283.3 401 Missense Mutation AAC,GAC N71D NP_444513.1

View Full Product Details