Product Details

SNP ID
rs32853
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:69033635 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTGAGATAAAATTGAACAGCTTCC[C/T]TTCTGAGAAATCACCAAACTGGAGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RPS2P45 PubMed Links

Gene Details

Gene
RPS2P45
Gene Name
ribosomal protein S2 pseudogene 45
There are no transcripts associated with this gene.

Gene
TANGO6
Gene Name
transport and golgi organization 6 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024562.1 Intron NP_078838.1
XM_011523327.2 Intron XP_011521629.1

View Full Product Details