Product Details
- SNP ID
-
rs1060651
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:154025194 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- ACCCTGAGAAGAGCTTTTCAAGCTG[C/T]ATTCCAGAAATGCAACATTAGAGTC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ARHGEF26-AS1
PubMed Links
Gene Details
- Gene
- ARHGEF26-AS1
- Gene Name
- ARHGEF26 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details