Product Details

SNP ID
rs3905734
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:109330526 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAGCTCTTCATGTTGCGCTTACCA[A/G]AGGCCTCTTGTGGAGATTCCACTGC
Phenotype
MIM: 614125
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DPPA4 PubMed Links

Gene Details

Gene
DPPA4
Gene Name
developmental pluripotency associated 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018189.3 732 Missense Mutation TCT,TTT S226F NP_060659.3
XM_011512954.2 732 Missense Mutation TCT,TTT S174F XP_011511256.1

View Full Product Details