Product Details
- SNP ID
-
rs1746479
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:131136155 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAATGTAAACAAAACCTAGTAAAAT[C/T]TCGATTTTATCTTATATCACAAATA
- Phenotype
-
MIM: 604693
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
AKAP7
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77095004] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- AKAP7
- Gene Name
- A-kinase anchoring protein 7
View Full Product Details