Product Details

SNP ID
rs936257
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:108386061 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAAATAAAATCCCTTCTTTGTTTT[C/T]ATCTGCCTATCCCTCAACTTGTCAG
Phenotype
MIM: 604371 MIM: 609929
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HHLA2 PubMed Links

Gene Details

Gene
HHLA2
Gene Name
HERV-H LTR-associating 2
There are no transcripts associated with this gene.

Gene
MYH15
Gene Name
myosin heavy chain 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014981.1 Intron NP_055796.1
XM_011512559.2 Intron XP_011510861.1
XM_017005922.1 Intron XP_016861411.1

View Full Product Details