Product Details

SNP ID
rs917535
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:141199794 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGAACTATGGAGAACCAAGGGACAC[A/G]CACTCAGCAGATAAGGCAAGTCCTG
Phenotype
MIM: 606336 MIM: 606337 MIM: 606338 MIM: 606335
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PCDHB10 PubMed Links

Gene Details

Gene
PCDHB10
Gene Name
protocadherin beta 10
There are no transcripts associated with this gene.

Gene
PCDHB11
Gene Name
protocadherin beta 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018931.2 20 Missense Mutation CAC,CGC H7R NP_061754.1
Gene
PCDHB12
Gene Name
protocadherin beta 12
There are no transcripts associated with this gene.

Gene
PCDHB9
Gene Name
protocadherin beta 9
There are no transcripts associated with this gene.

View Full Product Details