Product Details

SNP ID
rs706523
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:25964236 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTTAACACCAATAATATTTATTA[C/T]CCATTTATTTCACAGACTTGCTATA
Phenotype
MIM: 608231
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RASSF8 PubMed Links

Gene Details

Gene
RASSF8
Gene Name
Ras association domain family member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164746.1 Intron NP_001158218.1
NM_001164747.1 Intron NP_001158219.1
NM_001164748.1 Intron NP_001158220.1
NM_007211.4 Intron NP_009142.2
XM_006719027.2 Intron XP_006719090.1
XM_006719028.1 Intron XP_006719091.1
XM_017018746.1 Intron XP_016874235.1
Gene
RASSF8-AS1
Gene Name
RASSF8 atnisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details