Product Details

SNP ID
rs3846795
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:63280189 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGCATCTTCTAGTTTTAAAGGGAT[C/T]TCAGAAGGTTCCACTTGGTAAAAGT
Phenotype
MIM: 611470
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LGSN PubMed Links

Gene Details

Gene
LGSN
Gene Name
lengsin, lens protein with glutamine synthetase domain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143940.1 2513 UTR 3 NP_001137412.1
NM_016571.2 2513 Silent Mutation GAA,GAG E454E NP_057655.2
XM_011535889.2 2513 Silent Mutation GAA,GAG E535E XP_011534191.1
XM_011535892.2 2513 Silent Mutation GAA,GAG E493E XP_011534194.1
XM_017010929.1 2513 Silent Mutation GAA,GAG E535E XP_016866418.1
XM_017010930.1 2513 Silent Mutation GAA,GAG E535E XP_016866419.1
XM_017010931.1 2513 Silent Mutation GAA,GAG E430E XP_016866420.1

View Full Product Details