Product Details

SNP ID
rs10516
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:51352050 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTCTCAGGGATGTAGGCGGCCTCC[C/T]GCAGGGTTGAGTTGCAATGGGAACA
Phenotype
MIM: 130120 MIM: 605148
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CELA1 PubMed Links

Gene Details

Gene
CELA1
Gene Name
chymotrypsin like elastase family member 1
There are no transcripts associated with this gene.

Gene
GALNT6
Gene Name
polypeptide N-acetylgalactosaminyltransferase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007210.3 4734 UTR 3 NP_009141.2
XM_005268607.1 4734 UTR 3 XP_005268664.1
XM_006719214.2 4734 UTR 3 XP_006719277.1
XM_011537819.2 4734 UTR 3 XP_011536121.1
XM_011537822.2 4734 UTR 3 XP_011536124.1
XM_017018744.1 4734 UTR 3 XP_016874233.1
XM_017018745.1 4734 UTR 3 XP_016874234.1

View Full Product Details