Product Details

SNP ID
rs1468158
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.16:31704610 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ATTCTGAAAAGAAAGTCAGGCAGTA[C/T]GAAGCTAGGGCTTGGAATCCGCCCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CLUHP3 PubMed Links

Gene Details

Gene
CLUHP3
Gene Name
clustered mitochondria homolog pseudogene 3
There are no transcripts associated with this gene.

Gene
ZNF720
Gene Name
zinc finger protein 720
There are no transcripts associated with this gene.

View Full Product Details