Product Details

SNP ID
rs834179
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:42988162 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTCTGGCCTCCAGCCCAGCTGAC[C/T]ACCACTGCTGGAGAGTGGGTAGGGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM198A PubMed Links

Gene Details

Gene
FAM198A
Gene Name
family with sequence similarity 198 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001129908.2 Intron NP_001123380.2

View Full Product Details