Product Details

SNP ID
rs1803267
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:241678561 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGCCAAACGCTCCCCGCTTGGCA[G/T]CATCCGCCAGCTGTAACTGGAGGAA
Phenotype
MIM: 611338 MIM: 188345 MIM: 608525
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ATG4B PubMed Links

Gene Details

Gene
ATG4B
Gene Name
autophagy related 4B cysteine peptidase
There are no transcripts associated with this gene.

Gene
DTYMK
Gene Name
deoxythymidylate kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001165031.1 441 Missense Mutation GAT,GCT D116A NP_001158503.1
NM_001320902.1 441 Missense Mutation GAT,GCT D97A NP_001307831.1
NM_001320903.1 441 Missense Mutation ATG,CTG M86L NP_001307832.1
NM_001320904.1 441 Missense Mutation GAT,GCT D121A NP_001307833.1
NM_001320905.1 441 Missense Mutation GAT,GCT D179A NP_001307834.1
NM_012145.3 441 Missense Mutation GAT,GCT D140A NP_036277.2
Gene
ING5
Gene Name
inhibitor of growth family member 5
There are no transcripts associated with this gene.

View Full Product Details