Product Details

SNP ID
rs877052
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:78998148 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGTTGGTTCGGTCAATACCAAAGG[C/T]TCTTGTATGATGACTGCTGGGTAAT
Phenotype
MIM: 605849
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DMGDH PubMed Links

Gene Details

Gene
DMGDH
Gene Name
dimethylglycine dehydrogenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013391.3 2589 Silent Mutation GAA,GAG E845E NP_037523.2
XM_006714597.2 2589 Intron XP_006714660.1
XM_011543354.2 2589 Intron XP_011541656.1
XM_011543355.2 2589 Intron XP_011541657.1

View Full Product Details