Product Details
- SNP ID
-
rs1410331
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:79315191 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTTTACCAAAACCTCTCTGCATCAT[A/G]CTCCCTTCAAACATTAGGCTGCAAA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RBM26
PubMed Links
Gene Details
- Gene
- RBM26
- Gene Name
- RNA binding motif protein 26
View Full Product Details