Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001256264.1 | 3789 | Silent Mutation | CAC,CAT | H1248H | NP_001243193.1 |
NM_001256265.1 | 3789 | Silent Mutation | CAC,CAT | H1204H | NP_001243194.1 |
NM_002018.3 | 3789 | Silent Mutation | CAC,CAT | H1259H | NP_002009.1 |
XM_005256555.3 | 3789 | Silent Mutation | CAC,CAT | H1286H | XP_005256612.1 |
XM_005256556.3 | 3789 | Silent Mutation | CAC,CAT | H1285H | XP_005256613.1 |
XM_005256558.2 | 3789 | Silent Mutation | CAC,CAT | H1258H | XP_005256615.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_004140.3 | 3789 | Intron | NP_004131.3 | ||
XM_011523849.2 | 3789 | Intron | XP_011522151.1 | ||
XM_011523850.2 | 3789 | Intron | XP_011522152.1 | ||
XM_011523851.2 | 3789 | Intron | XP_011522153.1 | ||
XM_011523852.2 | 3789 | Intron | XP_011522154.1 | ||
XM_011523853.2 | 3789 | Intron | XP_011522155.1 | ||
XM_011523854.2 | 3789 | Intron | XP_011522156.1 | ||
XM_011523855.2 | 3789 | Intron | XP_011522157.1 | ||
XM_011523856.1 | 3789 | Intron | XP_011522158.1 |