Product Details

SNP ID
rs2018861
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:45632982 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTCACACACATAAGCAGGGAGAAGT[C/T]GTGAAGATACATAAACAGCTGCGTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CRHR1-IT1 PubMed Links

Gene Details

Gene
CRHR1-IT1
Gene Name
CRHR1 intronic transcript 1
There are no transcripts associated with this gene.

Gene
MGC57346
Gene Name
ADP-ribosylation factor pseudogene
There are no transcripts associated with this gene.

Gene
MGC57346-CRHR1
Gene Name
MGC57346-CRHR1 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256299.2 Intron NP_001243228.1
NM_001303016.1 Intron NP_001289945.1

View Full Product Details