Product Details

SNP ID
rs1042638
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:80037711 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTTAAGAGAACAATTTTAGTTAA[A/G]AATCAAATATACAGAGATTCAAAGA
Phenotype
MIM: 611990 MIM: 604068
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MRPS28 PubMed Links

Gene Details

Gene
MRPS28
Gene Name
mitochondrial ribosomal protein S28
There are no transcripts associated with this gene.

Gene
TPD52
Gene Name
tumor protein D52
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025252.2 1176 UTR 3 NP_001020423.1
NM_001025253.2 1176 UTR 3 NP_001020424.1
NM_001287140.1 1176 UTR 3 NP_001274069.1
NM_001287142.1 1176 UTR 3 NP_001274071.1
NM_001287143.1 1176 UTR 3 NP_001274072.1
NM_001287144.1 1176 UTR 3 NP_001274073.1
NM_005079.3 1176 UTR 3 NP_005070.1

View Full Product Details