Product Details

SNP ID
rs829164
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:37407384 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCGCAGGTAAAATGACTGGCCCAG[G/T]CCGGTCCCACCCCGTAAGGATTTGA
Phenotype
MIM: 200350 MIM: 602276
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ACACA PubMed Links

Gene Details

Gene
ACACA
Gene Name
acetyl-CoA carboxylase alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198834.2 160 Intron NP_942131.1
NM_198836.2 160 Intron NP_942133.1
NM_198837.1 160 Intron NP_942134.1
NM_198838.1 160 Intron NP_942135.1
NM_198839.2 160 Intron NP_942136.1
XM_005257267.4 160 Intron XP_005257324.1
XM_006721853.1 160 Intron XP_006721916.1
XM_011524701.1 160 Intron XP_011523003.1
XM_011524703.1 160 Intron XP_011523005.1
XM_011524704.2 160 Intron XP_011523006.1
XM_017024553.1 160 Intron XP_016880042.1
XM_017024554.1 160 Intron XP_016880043.1
XM_017024555.1 160 Intron XP_016880044.1
Gene
TADA2A
Gene Name
transcriptional adaptor 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166105.2 160 Intron NP_001159577.2
NM_001291918.1 160 Intron NP_001278847.1
NM_001488.4 160 UTR 5 NP_001479.4
NM_133439.3 160 UTR 5 NP_597683.3
XM_006722044.3 160 Intron XP_006722107.1
XM_011525153.2 160 Intron XP_011523455.1
XM_011525154.2 160 UTR 5 XP_011523456.1
XM_011525155.2 160 Intron XP_011523457.1
XM_017024981.1 160 Intron XP_016880470.1
XM_017024982.1 160 Intron XP_016880471.1
XM_017024983.1 160 Intron XP_016880472.1
XM_017024984.1 160 Intron XP_016880473.1
XM_017024985.1 160 Intron XP_016880474.1
XM_017024986.1 160 Intron XP_016880475.1

View Full Product Details