Product Details

SNP ID
rs2227338
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.17:40016689 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
TGCTGGCATTCTGGGCACCACAGCC[A/G]GGCCTGTGTATGGGCCCTGTCCATG
Phenotype
MIM: 138970 MIM: 607000
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CSF3 PubMed Links

Gene Details

Gene
CSF3
Gene Name
colony stimulating factor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000759.3 Intron NP_000750.1
NM_001178147.1 Intron NP_001171618.1
NM_172219.2 Intron NP_757373.1
NM_172220.2 Intron NP_757374.2
Gene
MED24
Gene Name
mediator complex subunit 24
There are no transcripts associated with this gene.

Gene
MIR6884
Gene Name
microRNA 6884
There are no transcripts associated with this gene.

View Full Product Details