Product Details

SNP ID
rs1370367
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:54359253 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GACAGAAAACATTTGGTCAGGCTTA[C/T]CCAAACAAATTTACTTGAGTGAGTT
Phenotype
MIM: 613258 MIM: 607051
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C18orf54 PubMed Links

Gene Details

Gene
C18orf54
Gene Name
chromosome 18 open reading frame 54
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288980.1 Intron NP_001275909.1
NM_001288981.1 Intron NP_001275910.1
NM_001288982.1 Intron NP_001275911.1
NM_173529.5 Intron NP_775800.3
XM_005258201.1 Intron XP_005258258.1
XM_005258203.1 Intron XP_005258260.1
XM_017025567.1 Intron XP_016881056.1
Gene
STARD6
Gene Name
StAR related lipid transfer domain containing 6
There are no transcripts associated with this gene.

View Full Product Details