Product Details

SNP ID
rs1044268
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:33177638 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAATGTCATTTAAAGTTAAGGCTT[C/T]GCTGTTCATTTTGAAACAACAATTT
Phenotype
MIM: 602069
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NRP1 PubMed Links

Gene Details

Gene
NRP1
Gene Name
neuropilin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024628.2 5402 Intron NP_001019799.1
NM_001024629.2 5402 Intron NP_001019800.1
NM_001244972.1 5402 UTR 3 NP_001231901.1
NM_001244973.1 5402 UTR 3 NP_001231902.1
NM_003873.5 5402 UTR 3 NP_003864.4
XM_006717521.1 5402 UTR 3 XP_006717584.1
XM_006717522.1 5402 UTR 3 XP_006717585.1
XM_006717523.2 5402 Intron XP_006717586.1
XM_006717524.1 5402 UTR 3 XP_006717587.1
XM_006717525.1 5402 UTR 3 XP_006717588.1
XM_006717526.1 5402 Intron XP_006717589.1
XM_011519755.2 5402 Intron XP_011518057.1
XM_011519756.1 5402 Intron XP_011518058.1
XM_017016865.1 5402 Intron XP_016872354.1
XM_017016866.1 5402 UTR 3 XP_016872355.1

View Full Product Details