Product Details

SNP ID
rs1056401
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.17:513484 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
TGTAGGCCAGCATTTCAGGTGTCCA[G/T]GTTTCTAAGGCTTTTGCTTAAATTG
Phenotype
MIM: 615850
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
VPS53 PubMed Links

Gene Details

Gene
VPS53
Gene Name
VPS53, GARP complex subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128159.2 8531 UTR 3 NP_001121631.1
NM_018289.3 8531 Intron NP_060759.2
XM_011523953.2 8531 Intron XP_011522255.1
XM_017024817.1 8531 UTR 3 XP_016880306.1
XM_017024818.1 8531 UTR 3 XP_016880307.1

View Full Product Details