Product Details

SNP ID
rs889548
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:31126391 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTCCAGAGATGTCAGAAGTAGACT[T/C]GACATCCACTGTATTAGGACTTGGC
Phenotype
MIM: 614901 MIM: 609912 MIM: 600823
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
BCKDK PubMed Links

Gene Details

Gene
BCKDK
Gene Name
branched chain ketoacid dehydrogenase kinase
There are no transcripts associated with this gene.

Gene
KAT8
Gene Name
lysine acetyltransferase 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032188.2 Intron NP_115564.2
NM_182958.2 Intron NP_892003.2
XM_011545970.1 Intron XP_011544272.1
XM_011545971.1 Intron XP_011544273.1
Gene
PRSS8
Gene Name
protease, serine 8
There are no transcripts associated with this gene.

View Full Product Details