Product Details

SNP ID
rs9853
Assay Type
Functionally tested
NCBI dbSNP Submissions
36
Location
Chr.1:160277198 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCAGACTTGTGCTGGTCAGTGAACA[A/C]CTTTTTTTTTTTTTCACCAGTCTGT
Phenotype
MIM: 600279
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PEX19 PubMed Links

Gene Details

Gene
PEX19
Gene Name
peroxisomal biogenesis factor 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193644.1 3280 UTR 3 NP_001180573.1
NM_002857.3 3280 UTR 3 NP_002848.1

View Full Product Details